Learn More
Cole-Parmer Cleanroom Panel-Mount Female Luer to Hose Barb Fittings, Straight Adapter, Nylon
Shop All Cole-Parmer ProductsDescription
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 1/16 in. |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Inlet | Female Luer, Hose Barb |
| Color | White |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.