Learn More
Cole-Parmer Cleanroom NPT(M) to Hose Barb Fittings, Straight Adapter, White Nylon
Product Code. 30053401 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Thread Size:
1/2 in. NPT(M)
1/4 in. NPT(M)
1/8 in. NPT(M)
3/8 in. NPT(M)
Unit Size:
Pack of 10
Product Code. 30053401
Brand: Cole-Parmer 5010785
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 5/16 in.
Packaging Quantity 10/Pk.
Outlet Port Threaded NPT(M) x Hose Barb
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Thread Size 1/4 in. NPT(M)
Shape Straight
Color White