Learn More
Cole-Parmer Cleanroom Male Slip Luer to Barb Fittings, Straight Adapter, Natural Kynar™
Product Code. 30053073 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/4 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
30053073 3/32 in. Pack of 10
30053252 1/4 in. Pack of 10
30054089 5/32 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30053073 Supplier Cole-Parmer Supplier No. 5011486
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;USP
Diameter (English) Inner 3/32 in.
Material Kynar
Packaging Quantity 10/Pk.
Inlet Male slip luer
Color Natural