Learn More
Cole-Parmer Cleanroom Male Luer Slip Plugs, Straight Adapter
Product Code. 30053263 Shop All Cole-Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
Product Code. 30053263
Brand: Cole-Parmer 5011058
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Seal luer ports with a secure and leak-proof connection. Ideal for capping off fluid transfer system. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Material Nylon
Inlet Male Luer Slip, Plug
Color Natural