Learn More
Cole-Parmer Cleanroom Male Luer Slip Plugs, Straight Adapter
Shop All Cole-Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
Description
Seal luer ports with a secure and leak-proof connection. Ideal for capping off fluid transfer system. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Inlet | Male Luer Slip, Plug |
| Color | Natural |