Learn More
Cole-Parmer Cleanroom Large-Bore Male Luer to Hose Barb Fittings, Straight Adapter, Natural Kynar™
Alle ansehen Cole-Parmer Produkte
missing translation for 'orderingAttributeHoverText'
Diameter (English) Inner:
1/4 in.
3/16 in.
5/16 in.
missing translation for 'unitSize'
Pack of 10
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Spezifikation
Spezifikation
| Certifications/Compliance | Non-DEHP;Non-Phthalate;USP |
| Diameter (English) Inner | 1/4 in. |
| Material | Kynar |
| Packaging Quantity | 10/Pk. |
| Inlet | Large Bore Male Luer, Hose Barb |
| Color | Natural |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur