Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer to Hose Barb Fittings, Straight Adapter, Natural Kynar™
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/4 in.
3/16 in.
5/16 in.
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;USP |
| Diameter (English) Inner | 1/4 in. |
| Material | Kynar |
| Packaging Quantity | 10/Pk. |
| Inlet | Large Bore Female Luer, Hose Barb |
| Color | Natural |