Learn More
Cole-Parmer Cleanroom Female Luer Plugs, Straight Adapter
Shop All Cole-Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Inlet:
Female Luer with Tether
Female Luer, Plug
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Inlet | Female Luer, Plug |
| Color | Natural |