Learn More
Cole-Parmer Cleanroom Female Luer Plugs, Straight Adapter
Product Code. 30053051 Shop All Cole-Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Inlet:
Female Luer with Tether
Female Luer, Plug
Unit Size:
Pack of 10
Product Code. 30053051
Brand: Cole-Parmer 5011001
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Material Polypropylene
Packaging Quantity 10/Pk.
Inlet Female Luer with Tether
Color Clear