missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Branson Ultrasonics™ Cleaning Reagent, Industrial Strength
Description
- Heavier greases, oils, and particulates
- Drawing lubricants
- Light carbon deposits
- Removal of heavier soils from automotive, aircraft, and similar mechanical components
- Cleaning prior to secondary finishing operations (including painting, plating, and electrostatic coatings)
Specifications
Specifications
| Product Type | Cleaning Reagent |
| pH | 13 |
| For Use With (Application) | For Industrial |
| Volume (Metric) | 1 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?