Learn More
Hirschmann™ Cleaning Detergent rea-clean™
Product Code. p-7162677
Click to view available options
Volume (Metric):
1 L
30 L
Unit Size:
Each
Product Code. 10160661
Brand: Hirschmann™ 9790101
Bio degradable cleaning detergent for rapid, automatic and careful cleaning of materials and surfaces especially made of plastics.
  • Biodegradable cleaner
  • Surface cleaning
  • Phosphate-free and self-neutralising
Specifications
Product Type Phosphate-free Cleaning Concentrate
Form Liquid
Product Line rea-clean
Description Biodegradable, fat dissolving, gentle on materials
For Use With (Equipment) All sufaces in labs
Quantity 1 Refill Bottle
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • rea-clean Bio-Reiniger
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.