Learn More
Sampling Systems™ Cleaning Coupons for Metal
Product Code. p-7146958 Shop All Sampling Systems Products
Click to view available options
Material:
Aluminum
Stainless Steel (304)
Stainless Steel (316)
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
50 x 50 mm
Finish:
Bright Polish
Satin finish
missing translation for 'unitSize'
Each
Product Code. 15889541
missing translation for 'mfr': Sampling Systems™ 8644AL50
Only null left
Add to basket
Add to basket
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product Type Cleaning Coupon
Dimensions (L x W) 50 x 50 mm
Finish Bright Polish
For Use With (Application) Testing and validating cleaning regime
Material Aluminum