Learn More
ZVG Zellstoff Vertrieb™ Multisoft™ Cleaning Cloths
Product Code. p-8010369
Click to view available options
Dimensions (L x W):
37 x 38 cm
38 x 22 cm
Ply:
2-ply
4-ply
Unit Size:
1 roll
2 rolls
Product Code. 10341342
Brand: ZVG Zellstoff Vertrieb™ 1142500
Sensitive surfaces must be treated softly with ZVG Zellstoff Vertrieb™ Multisoft™ Cleaning Cloths. The extra-soft, lint-free special towels are the first choice when it comes to care and polishing. They absorb liquids such as polishing wax and release it evenly without losing their strength.
  • Very soft
  • White
  • 2-ply
Specifications
Product Type Cleaning Cloth
Color White
Dimensions (L x W) 38 x 22 cm
Format Roll
Packaging Quantity 30 Rolls
Ply 2-ply