Learn More
Phenomenex™ Cleanert™ Screw Caps, No Hole, with Septa, 22 mm, No Slit
Product Code. 31745354
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31745354 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31745354 Supplier Phenomenex™ Supplier No. AV79200
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Accessories for use with chromatography vials and in various chromatography applications.
  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Color White
Diameter (Metric) Outer 24 mm
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 24 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color Natural