Learn More
Phenomenex™ Cleanert™ Insert Vials, Glass, 6 mm
Product Code. 31746857 Shop All Phenomenex Products
Change view
Click to view available options
Insert Type:
Flat Bottom
V-Bottom
V-Bottom with Spring
Volume (Metric) Insert:
150 μL
200 μL
250 μL
300 μL
Unit Size:
Each
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Insert Type Volume (Metric) Insert Unit Size
31746857 V-Bottom 200 μL Each
31742527 V-Bottom with Spring 150 μL Each
31743342 Flat Bottom 300 μL Each
31741830 V-Bottom with Spring 250 μL -
31745690 V-Bottom 250 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31746857 Supplier Phenomenex™ Supplier No. AV10326
Estimated Shipment: 23-11-2026
Accessories for use with chromatography vials and in various chromatography applications.
  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 31 mm
Material Glass
Pre-slit No
Sterility Sterile
Insert Type V-Bottom
Volume (Metric) Insert 200 μL
Closure Size 6 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Insert