Learn More
Emma Safety Footwear Clay S3 Shoe
Product Code. 17592626
Change view
Click to view available options
Size:
39
40
41
42
45
46
47
48
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17592626 40 1 pair
17582626 39 1 pair
17502636 41 1 pair
17512636 42 1 pair
17522636 45 1 pair
17532636 46 1 pair
17542636 47 1 pair
17552636 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17592626 Supplier Emma Safety Footwear Supplier No. MM98054940
Only null left
Add to basket
Add to basket
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.
  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 40
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe