Learn More
Cytiva Classical Powdered Ham's Nutrient Mixture F10 - F12
Product Code. 11948075
Change view
Click to view available options
Quantity:
2 x 5 L
1 x 10 L
1 x 50 L
Unit Size:
10 L
50 L
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11948075 2 x 5 L Pack of 2
11561791 1 x 10 L 10 L
10733374 1 x 50 L 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11948075 Supplier Cytiva Supplier No. SH30010.02
  • ONLINE EXCLUSIVE PRICE
    27.42€
    33.80€ / Pack of 2
    Save POA
    -23.27% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Ham's F10-F12 Nutrient Mixture, manufactured using ISO 9001:2000-certified processes.
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Product Type Ham's F12, with L-glutamine
Sterilization Method Non-sterile
With Additives Glutamine
Without Additives No Sodium Bicarbonate, Sodium Bicarbonate
Quantity 2 x 5 L
Certifications/Compliance cGMP and ISO 9001:2000
Content And Storage 2° to 8°C, away from light
Description Ham's F12, with L-Glutamine