Learn More
Value Plastics Classic Barb to Press-In Plug Fitting, Nylon
Shop All Cole-Parmer Products
Click to view available options
Material:
Nylon
Unit Size:
Pack of 25
Description
Ideal for capping off fluid transfer system. Constructed of durable chemical resistance nylon. Classic barb style offers greater sealing capability
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Inlet | Plug, Classic Hose Barb |
| Outlet Port | Hose Barb x Plug |
| Sterilization Method | Gamma radiation, ethylene oxide (EtO), and autoclave |
| Shape | Plug |