Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Click to view available options
Mass:
1 g
1 to 100 g
10 g
100 g
2 g
20 g
200 g
5 g
5 kg
50 g
Pakningsstørrelse:
Each
Beskrivelse
- Class E2 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Tekniske data
Tekniske data
| Mass | 10 g |
| Tolerance | ±0.06 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion