Learn More
VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale
Product Code. p-8003651
Change view
Click to view available options
Graduations:
0.2 mL
0.5 mL
1 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 12
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Graduations unitSize
10471525 0.2 mL Pack of 12
10028630 20 mL Each
11837930 0.5 mL Pack of 12
11827930 5 mL Pack of 6
10782637 1 mL Pack of 12
15782106 10 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10471525 Supplier VITLAB™ Supplier No. 646081
Add to basket
Add to basket
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale feature easy to read, raised, embossed blue scale and ring marks at the primary scale points.
  • Highly transparent polypropylene graduated cylinder
  • Hexagonal base with bottom studs provides high stability
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
  • To preserve markings, do not clean at temperatures exceeding 60°C
Specifications
Tolerance ±0.2 mL
Class Class B
Quantity 6/Pk.
Graduations 0.2 mL
Limit of Error ±0.2mL
Shape Round
For Use With (Application) Food Industry and Volume Measurement
Scale Color Blue
Base Type Hexagonal
Height (Metric) 145 mm
Product Type Graduated Cylinder