Learn More
VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale
Description
- Highly transparent polypropylene graduated cylinder
- Hexagonal base with bottom studs provides high stability
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
- To preserve markings, do not clean at temperatures exceeding 60°C
Specifications
Specifications
| Material | Polypropylene |
| Tolerance | ±1 mL |
| Certifications/Compliance | DIN 12681, ISO 6706 |
| Class | Class B |
| Diameter (Metric) | 27 mm |
| Quantity | 6/Pk. |
| Graduations | 1 mL |
| Limit of Error | ±1mL |
| Shape | Round |
| For Use With (Application) | Food Industry and Volume Measurement |
| Show More |