Learn More
Hirschmann™ Class B Measuring Cylinders, Blue Stain Graduation
Product Code. 11768021
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 11768021
Brand: Hirschmann™ 2230180
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Blue Stain Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, short line
  • Blue stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Base Type Hexagonal
Scale Color Blue
Shape Round
Certifications/Compliance DIN EN ISO 4788
Class B
Height (Metric) 260 mm
Tolerance Metric ±0.75 mL
Type Graduated Measuring Cylinder
Material Glass
Capacity (Metric) 100 mL
Graduations 1 mL
Limit of Error 0.75