Learn More
Brand™ Class B AR-Glass/Soda Lime Glass SilberEterna™ Volumetric Bulb Pipet
Product Code. 11878271 Shop All Brand Products
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11878271
Brand: Brand™ 29518
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These bulb pipettes provide the highest accuracy, maintained by strict statistical quality controls.
  • Acc. to DIN EN ISO 648
  • 0.5 to 2mL capacity bulbs provided in a pack of 12
  • 3 to 100mL bulbs provided in a pack of 6
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Calibrated Yes
Certifications/Compliance DIN EN ISO 648
Class B
Material AR-GLAS, Soda Lime Glass
Graduated Yes
Graduation Color Amber
Quantity 6 Pack
Tolerance ±0.07 mL
Volume (Metric) 50 mL
Length (Metric) 540 mm
Type Volumetric Bulb Pipette