Learn More
Brand™ Class B AR-Glass/Soda Lime Glass SilberEterna™ Volumetric Bulb Pipet
Shop All Brand Products
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Description
- Acc. to DIN EN ISO 648
- 0.5 to 2mL capacity bulbs provided in a pack of 12
- 3 to 100mL bulbs provided in a pack of 6
Specifications
Specifications
| Autoclavable | Autoclavable |
| Calibrated | Yes |
| Certifications/Compliance | DIN EN ISO 648 |
| Class | B |
| Material | AR-GLAS, Soda Lime Glass |
| Graduated | Yes |
| Graduation Color | Amber |
| Quantity | 12 Pack |
| Tolerance | ±0.010 mL |
| Volume (Metric) | 1 mL |
| Show More |