Learn More
Hirschmann™ Class A Measuring Cylinders, Amber Graduation
Product Code. p-8011832
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 10542404
Brand: Hirschmann™ 2210154
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Amber Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, main point ring amber stain graduation
  • Conformity certified, with dated batch identification
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Base Type Hexagonal
Calibration To Contain
Description Calibration To Contain
Scale Color Brown
Shape Round
Sterility Non-sterile
Certifications/Compliance DIN EN ISO 4788
Class A
Height (Metric) 115 mm
Tolerance Metric ±0.05 mL
Type Graduated Measuring Cylinder
Material Glass
Capacity (Metric) 5 mL
Graduations 0.1 mL
Autoclavable Autoclavable
Subdivision 1 mL