Learn More
Afora™ Class A Graduated Pipet
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
Each
Specifications
Specifications
| Class | A |
| Graduated | Yes |
| Graduations | 1 mL |
| Quantity | 1 Pack |
| Volume (Metric) | 10 mL |
| Type | Class A Pipette |