Learn More
Afora™ Class A Graduated Pipet
Brand:  Afora™ A6002
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12004618
  • 29.90€ / Each
Estimated Shipment: 22-05-2024
Specifications
A
1 mL
5 mL
Yes
1 Pack
Class A Pipette