Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Shop All Brand Products
Click to view available options
Socket Joint:
NS 12/21
NS 14/23
Unit Size:
Each
Pack of 2
Description
- Borosilicate 3.3
- DIN EN ISO 1042
- Calibrated to contain (TC, In)
- Includes ISO individual certificate
Specifications
Specifications
| Certifications/Compliance | ISO Individual |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass (3.3) |
| Socket Joint | NS 14/23 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Mouth Style | Narrow |
| Sterility | Non-sterile |
| Bottom Shape | Flat |