Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
The A1P3 cartridge is part of the wide range of cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications. The range provides high quality and a cost-effective protection.
Brand: Honeywell™ N06575081L
Additional Details : Weight : 0.09000kg
Description
- Protects against organic vapor and particulates
- Excellent filtration efficiency over extensive period of time
- Lower profile and lightweight for optimal comfort
- Low profile does not obstruct the field of vision
- Threaded connector is easy to attach
Specifications
Class 1 plastic cartridge | |
Protects against organic vapor and particulates | |
92.5 g |
EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259 | |
6 Pair/Box, 8 Boxes/Cs. |