Learn More
Medline Scientific™ Clamp Flask
Product Code. 10644133
Change view
Click to view available options
Holds:
1 L Flask
2 L Flask
50 mL Flask
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10644133 1 L Flask Each
10589712 50 mL Flask Each
10225522 2 L Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10644133 Supplier Medline Scientific™ Supplier No. AAA23554
  • ONLINE EXCLUSIVE PRICE
    60.90€
    55.96€ / Pack of 1
    Save 4.94€
    8% Off
Estimated Shipment: 20-01-2027
For use with bench top and floor standing shaking incubators
The Lab Companion range of refrigerated shaking incubators include the SI-600R, a smaller version, the SI-300R and the floor model IS-971R which is a capable of incubating large volumes and very low temperatures (+4°C).
The SI-300R and SI-600R units are bench top models with a slim width to ensure they fit into the tightest of spaces. The lid, which is supported by integral gas pressure struts, opens to give front and top access for any size flask up to 2,000mL. The patented CLS control system exceeds Class II safety standards for use in areas where there may be flammable environments.
PID controller ensures accurate temperature control
  • LED display
  • Select orbital or reciprocal motion
  • Cooling coils supplied with non-refrigerated models
  • Adjustable stroke length
  • Supplied with universal platform
Range of bench top and floorstanding shaking incubators suitable for a variety of applications.
Specifications
Holds 1 L Flask
Product Type Clamp Flask