Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Shop All Heidolph Instruments Products
Click to view available options
Holds:
100 mL Erlenmeyer flasks
1000 mL Erlenmeyer flasks
2000 mL Erlenmeyer flasks
25 mL Erlenmeyer flasks
250 mL Erlenmeyer flasks
50 mL Erlenmeyer flasks
500 mL Erlenmeyer flasks
Unit Size:
Each
Description
- Maximum number of clamps per platform type:
- 549-51000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-52000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-55000-00
- For platform 100 - max. 3
- For platform 1000 - max. 4
- For platform 2000 - max. 9
- 549-56000-00
- For platform 100 - max. 2
- For platform 1000 - max. 4
- For platform 2000 - max. 5
- 549-63000-00
- For platform 1000 - max. 2
- For platform 2000 - max. 3
- 549-51000-00
Specifications
Specifications
| Holds | 2000 mL Erlenmeyer flasks |
| Product Type | Clamp |
| For Use With (Equipment) | Perforated platforms 1000 and 2000 |
| Size | 7 |