Learn More
Lenz Laborglasintrumente™ Claisen Distilling Links with Vacuum Receiver Adapter
Click to view available options
Standard Taper Joint:
Cone NS 14/23
Unit Size:
Each
Description
- According to DIN 12594
- With Liebig condenser (angled 75°) and vertical Claisen head
- Water and vacuum connection
- With two sockets NS 14/23 for distillation thermometers of 55 mm built-in length
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 250 mm |
| Type | Claisen Distilling link |
| Material | DURAN™ tubing |
| Quantity | 1 |
| Standard Taper Joint | Cone NS 14/23 |