Learn More
Citric Acid Monohydrate, for Foodstuffs Analysis, low in lead, Fisher Chemical
Product Code. 10776091 Shop All Fisher Chemical Products
Change view
Click to view available options
:
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10776091 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10776091 Supplier Fisher Chemical Supplier No. C/6180/53
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
pH 1.8
CAS 5949-29-1
Melting Point 135°C
Color White
Molecular Formula C6H10O8
MDL Number 149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular Weight (g/mol) 210.14
Formula Weight 210.15g/mol
Quantity 500 g
Packaging Plastic powder jar
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.