Learn More
Citric Acid-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | Citric Acid-13C6 |
| CAS | 287389-42-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13C6 H8 O7 |
| Quantity | 5 mg |
| Synonym | Citric Acid-13C6, 99 atom % 13C, 2-Hydroxy-1,2,3-Propane-1,2,3-13C3-tri(carboxylic-13C) Acid |
| InChI Formula | InChI=1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1+1,2+1,3+1,4+1,5+1,6+1 |
| SMILES | O[13C](=O)[13CH2][13C](O)([13CH2][13C](=O)O)[13C](=O)O |
| IUPAC Name | 2-hydroxy(1,2,3-^{13}C_{3})propane-1,2,3-tricarboxylic acid |
| Molecular Weight (g/mol) | 198.08 |
| Show More |