missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CITFA
GREENER_CHOICE

Product Code. 18756376
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18756376

Brand: Tocris Bioscience™ 7811/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent G protein-coupled estrogen receptor (GPER) agonist

CITFA is a potent GPER agonist (EC50 = 38.7 nM). It significantly increases axonal and dendritic growth in rat embryonic (E18) hippocampal neurons.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-Cyclohexyl-4-(1-methylethyl)phenyl]-5-(tetrahydro-2H-pyran-2-yl)-2-furanmethanamine
CAS 2379989-52-1
Quantity 10 mg
Target GPER/GPR30
Molecular Formula C25H35NO2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.