missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-cis,trans-Abscisic Acid-d6, TRC
Specifications
Specifications
| Chemical Name or Material | (+)-cis,trans-Abscisic Acid d6 ((2-methyl-d3)cyclohex-2-en-1-yl-3,5,5-d3) |
| CAS | 721948-65-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H6 H14 O4 |
| Quantity | 1 mg |
| Synonym | 2,4-Pentadienoic acid, 5-[(1S)-1-hydroxy-6,6-dimethyl-2-(methyl-d3)-4-oxo-2-cyclohexen-1-yl-3,5,5-d3]-3-methyl-, (2Z,4E)-, (2Z,4E)-5-[(1S)-1-Hydroxy-6,6-dimethyl-2-(methyl-d3)-4-oxo-2-cyclohexen-1-yl-3,5,5-d3]-3-methyl-2,4-pentadienoic acid, [2H6](+)-cis,trans-Abscisic acid, (+)-cis,trans-Abscisic acid-d6, (+)-cis,trans-Abscisic acid D6 ((2-methyl-D3)cyclohex-2-en-1-yl-3,5,5-D3) |
| InChI Formula | InChI=1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m1/s1/i2D3,8D,9D2 |
| SMILES | [2H]C1=C(C([2H])([2H])[2H])[C@](O)(\C=C\C(=C/C(=O)O)\C)C(C)(C)C([2H])([2H])C1=O |
| IUPAC Name | (2Z,4E)-3-methyl-5-[(1S)-3,5,5-trideuterio-1-hydroxy-6,6-dimethyl-4-oxo-2-(trideuteriomethyl)cyclohex-2-en-1-yl]penta-2,4-dienoic acid |
| Molecular Weight (g/mol) | 270.35 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?