Learn More
cis-Tetrahydro-2,5(1H,3H)-pentalenedione, TRC
Specifications
Specifications
| Chemical Name or Material | cis-Tetrahydro-2,5(1H,3H)-pentalenedione |
| CAS | 51716-63-3 |
| Molecular Formula | C8H10O2 |
| Quantity | 1 g |
| Synonym | cis-Bicyclo[3.3.0]octane-3,7-dione |
| InChI Formula | InChI=1S/C8H10O2/c9-7-1-5-2-8(10)4-6(5)3-7/h5-6H,1-4H2/t5-,6+ |
| SMILES | O=C1C[C@@H]2CC(=O)C[C@@H]2C1 |
| IUPAC Name | 1,3,3a,4,6,6a-hexahydropentalene-2,5-dione |
| Molecular Weight (g/mol) | 138.16 |
| Formula Weight | 138.0681 |