Learn More
Tocris Bioscience™ cis-SIM1
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
cis-SIM1 is the negative control for SIM1 (Cat. No. 7432).
Specifications
Specifications
| Chemical Name or Material | N,N-(11-((2-(((S)-1-((2S,4S)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-2-oxoethoxy)methyl)-11-methyl-3,6,9,13,16,19-hexaoxahenicosane-1,21-diyl)bis(2-((S)-4-(4-chlorophenyl)-2,3,9-tri |
| CAS | 2719051-98-4 |
| Quantity | 2 mg |
| Target | Active Degrader Negative Controls |
| Molecular Formula | C79H98Cl2N14O13S3 |
| Purity | 98% |