Learn More
cis-Heptachlor Epoxide, TRC
Product Code. 30399113
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399113 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30399113 Supplier Toronto Research Chemicals Supplier No. TRCH26570010MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Heptachlor-exo-epoxide (cis-, isomer B)
CAS 1024-57-3
Molecular Formula C10 H5 Cl7 O
Quantity 10 mg
Synonym (1aR,1bS,2R,5S,5aR,6S,6aR)-rel-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, (1aα,1bβ,2α,5α,5aβ,6β,6aα)-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, 1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-3a,4,7,7a-tetrahydro-4,7-methanoindan, (±)-cis-Heptachlor epoxide, ENT 25584, Epoxyheptachlor, GPKh epoxide, HCE, HE, Hepox, Heptachlor cis-epoxide, Heptachlor epoxide, Heptachlor epoxide B, Heptachlor exo-epoxide, Heptepoxide, Velsicol 53CS17, cis-Heptachlor epoxide
InChI Formula InChI=1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8+,9-/m0/s1
SMILES Cl[C@H]1[C@H]2O[C@H]2[C@H]3[C@@H]1[C@@]4(Cl)C(=C(Cl)[C@]3(Cl)C4(Cl)Cl)Cl
Molecular Weight (g/mol) 389.32
Formula Weight 385.816