missing translation for 'onlineSavingsMsg'
Learn More

cis-Dimethylacetildenafil, TRC

Product Code. 30349310
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349310 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349310 Supplier Toronto Research Chemicals Supplier No. TRCD28212010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Des-N-Ethyl 3,5-Dimethylacetildenafil
CAS 1417999-76-8
Purity Grade Notes HPLC
Molecular Formula C25 H34 N6 O3
Quantity 10 mg
Synonym (3R,5S)-Dimethylacetildenafil
InChI Formula InChI=1S/C25H34N6O3/c1-6-8-19-22-23(30(5)29-19)25(33)28-24(27-22)18-11-17(9-10-21(18)34-7-2)20(32)14-31-12-15(3)26-16(4)13-31/h9-11,15-16,26H,6-8,12-14H2,1-5H3,(H,27,28,33)/t15-,16+
SMILES C(CC)C=1C2=C(C(=O)N=C(N2)C3=C(OCC)C=CC(C(CN4C[C@@H](C)N[C@@H](C)C4)=O)=C3)N(C)N1
IUPAC Name 5-(5-(2-((3R,5S)-3,5-dimethylpiperazin-1-yl)acetyl)-2-ethoxyphenyl)-1-methyl-3-propyl-1,4-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol) 466.58
Formula Weight 466.2692
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.