Learn More
cis-Chrysanthemic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | cis-Chrysanthemic Acid |
| CAS | 2935-23-1 |
| Molecular Formula | C10 H16 O2 |
| Quantity | 250 mg |
| Synonym | Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (1R,3S)-rel-, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (1R,3S)-rel- (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, cis-(±)-, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, cis-(±)- (8CI), rel-(1R,3S)-2,2-Dimethyl-3-(2-methyl-1-propen-1-yl)cyclopropanecarboxylic acid, (±)-cis-Chrysanthemic acid, (±)-cis-Chrysanthemumic acid, (±)-cis-Chrysanthemummonocarboxylic acid, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, cis-, DL-cis-Chrysanthemic acid, DL-cis-Chrysanthemumic acid, cis-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, cis-Chrysanthemic acid, cis-Chrysanthemumic acid, cis-DL-Chrysanthemumic acid, dl-cis-Chrysanthemic acid, dl-cis-Chrysanthemummonocarboxylic acid |
| InChI Formula | InChI=1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12)/t7-,8-/m1/s1 |
| SMILES | CC(=C[C@@H]1[C@H](C(=O)O)C1(C)C)C |
| IUPAC Name | (1S,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid |
| Molecular Weight (g/mol) | 168.233 |
| Formula Weight | 168.115 |