missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 15437046
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437046 250 mg 250mg
15417046 1 g 1g
15427046 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437046 Supplier Thermo Scientific Alfa Aesar Supplier No. H62109.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67299-52-9
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD01550737,MFCD01880738
InChI Key ZVMICQYOGWAOSU-UHFFFAOYSA-N
Synonym trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
PubChem CID 285247
SMILES OC(=O)C1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Solubility Information Soluble in water.
IUPAC Name 4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
Quantity 250 mg
Formula Weight 277.31
Percent Purity 97%
Chemical Name or Material cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.