missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Boc-amino)cyclobutanemethanol, 97%, Thermo Scientific™

Product Code. p-7041582
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429178 250 mg 250mg
15419178 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429178 Supplier Thermo Scientific Alfa Aesar Supplier No. H62686.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142733-64-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD08436074
InChI Key PCPNTJQMXAHNOA-UHFFFAOYSA-N
Synonym tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol
PubChem CID 22408683
IUPAC Name tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CC(C1)CO

Specifications

Quantity 250 mg
Formula Weight 201.26
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclobutanemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.