Learn More
cis-1,3,3-Trichloro-1-propene (cis-trans mixture, ~70% Z), TRC
Specifications
Specifications
| Chemical Name or Material | cis-1,3,3-Trichloro-1-propene |
| CAS | 2953-50-6 |
| Molecular Formula | C3H3Cl3 |
| Quantity | 10 mg |
| Synonym | (Z)-1,3,3-Trichloro-1-propene |
| InChI Formula | InChI=1S/C3H3Cl3/c4-2-1-3(5)6/h1-3H/b2-1- |
| SMILES | Cl\C=C/C(Cl)Cl |
| IUPAC Name | (Z)-1,3,3-trichloroprop-1-ene |
| Molecular Weight (g/mol) | 145.41 |
| Formula Weight | 143.93 |