Learn More
Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync)
Product Code. 10489391 Shop All Thermo Scientific Products
Click to view available options
Capacity (English):
4-place Distributor
8-place Distributor
Unit Size:
Each
Product Code. 10489391
Brand: Thermo Scientific™ 50091720
Only null left
Add to basket
Add to basket
Ensure identical operating conditions for multiple stirrer installations by using these benchtop distributors, for use with Mini, Micro, Compact and Maxi series stirrers.
Ensure identical operating conditions for multiple stirrer installations by using Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync), for use with Mini, Micro, Compact and Maxi series stirrers.
  • Simultaneous control of 2, 4, 6, or 8 Mini, Micro or Compact series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Simultaneous control of 2 or 4 Maxi Series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Even-powered distribution for synchronized stirring
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 4-place Distributor
For Use With (Equipment) Mini, Micro, Compact, and Maxi Series Stirrer
Type Benchtop Distributor