Learn More
Thermo Scientific™ Cimarec™ Biosystem 40B Controller
Shop All Thermo Scientific Products
Click to view available options
Voltage:
230 V Denmark
230 V Germany
230 V Switzerland
Unit Size:
Each
Description
Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.
- High visibility display
- Adjustable stirring and pause times with rotation sense inversion
Specifications
Specifications
| Description | Biomodul 40B Controller 230V Germany |
| Model | Biomodul 40B Controller |
| Type | Stirrer Controller (Dual) |
| Voltage | 230 V Germany |
| Electrical Requirements | 230 V |