Learn More
Thermo Scientific™ Cimarec™ 40M Controllers
Product Code. 10704921 Shop All Thermo Scientific Products
Click to view available options
Model:
Telemodul 40 M Control Units (40w)
Telemodul 80 M Control Units (80w)
Voltage:
115 V US
230 V Denmark
230 V Germany
230 V Swiss
230 V UK
Unit Size:
Each
Product Code. 10704921
Brand: Thermo Scientific™ 50118901
Maintain constant speeds, even during viscosity changes, with our 40M controllers.
Maintain constant speeds, even during viscosity changes, with Thermo Scientific™ Cimarec™ 40M Controllers. This product is required for use with the Thermo Scientific™ Cimarec™ Mobil Direct.
Features:
  • Adjustable speeds from 100 to 1000rpm
  • High visibility digital display
  • Adjustable start-up times
  • Thermal and electrical overload protection, with fault alert
Telemodul 40 M
  • Powerful controller equips Mobil 200 and 600 stirrers for the most demanding mixing tasks
Telemodul 80 M
  • Offers all the functionality of the Telemodul 40 M, with power output boosted to 80W for more demanding stirring tasks
TRUSTED_SUSTAINABILITY
Specifications
Model Telemodul 40 M Control Units (40w)
Speed 100 to 1000 rpm
Type Stirrer Controller
Voltage 230 V Germany
Electrical Requirements 230 V
Stirring Range 100 RPM to 1000 RPM
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.