Learn More
R&D Systems™ Ciliobrevin A
Description
Ciliobrevin A is a Hedgehog (Hh) pathway antagonist; blocks Sonic hedgehog (Shh)-induced Hh pathway activation (IC50 = 7 μM) downstream of Smo. Perturbs primary cilia formation; inhibits cytoplasmic AAA+ ATPase dynein-dependent microtubule gliding and ATPase activity.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-α-(3,4-4-oxo-2(1H)-quinazolinylidene)-β-oxobenzenepropanenitrile |
| CAS | 302803-72-1 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C17H9Cl2N3O2 |
| Formula Weight | Observed MW: 358.18 |