Learn More
Nickel Electro™ Churchill Test Tube Rack
Product Code. 12892294 Shop All Nickel-Electro Products
Click to view available options
Positions:
28
Unit Size:
Each
Product Code. 12892294
Brand: Nickel Electro™ C184
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from nylon offering good resistance to solvents, formaldehyde and alcohols with limited resistance to dilute acids. Each Churchill Test Tube Rack fits into Clifton water baths and can be easily removed and stored flat.
  • Churchill test tube racks have been developed in conjunction with a secondary school for a classroom of students to each place a test tube into the rack for incubation and other experiments in a water baths.
  • Churchill racks can accommodate the popular 18mm or 25mm diameter test tubes.
Specifications
Product Type Churchill Test Tube Rack
Description Churchill test tube rack 28 holes x 18 mm dia
Autoclavable Not Autoclavable
Color Gray
Holds Tubes (18 mm)
Material Nylon
Positions 28
Shape Rectangle
Sterility Non-sterile
For Use With (Equipment) Clifton Unstirred Water Baths 2 and 4 Litre Capacity
No. per Pack 1 Ea.
Quantity 1 Ea.
Dimensions (L x W x H) 350 x 200 x 20 mm
Diameter (Metric) Opening 18 mm