Learn More
Proteintech ChromoTek V5-Trap™ Agarose
Description
- Consists of an anti-V5 Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |